Läs mer
Deoxypyridinoline Chloride Trihydrochloride Salt, TRC
Produktkod. 30350714
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30350714 1 mg
1 options
Produktkod. 30350714 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2499001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Deoxypyridinoline chloride trihydrochloride
CAS 204074-56-6
Molekylformel C18 H28 N4 O7 . H Cl
Kvantitet 1 mg
Synonym 4-[(2S)-2-Amino-2-carboxyethyl]-1-[(5S)-5-amino-5-carboxypentyl]-3-[(3S)-3-amino-3-carboxypropyl]-5-hydroxypyridinium Chloride Trihydrochloride Salt, Lysylpyridinoline
InChI formel InChI=1S/C18H28N4O7.ClH/c19-12(16(24)25)3-1-2-6-22-8-10(4-5-13(20)17(26)27)11(15(23)9-22)7-14(21)18(28)29;/h8-9,12-14H,1-7,19-21H2,(H3-,23,24,25,26,27,28,29);1H/t12-,13-,14-;/m0./s1
LEDER Cl.N[C@@H](CCCC[n+]1cc(O)c(C[C@H](N)C(=O)[O-])c(CC[C@H](N)C(=O)O)c1)C(=O)O
IUPAC-namn (2S)-2-amino-3-[1-[(5S)-5-amino-5-carboxypentyl]-3-[(3S)-3-amino-3-carboxypropyl]-5-hydroxypyridin-1-ium-4-yl]propanoate;hydrochloride
Molekylvikt (g/mol) 448.899
Formel vikt 448.172