missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Deoxyribonuclease I (Bovine)
Used for Enzyme
Brand: Abnova™ P5307.25000Units
Additional Details : Weight : 0.00010kg
Description
#REF!Specifications
P00639 | |
282217 | |
Deoxyribonuclease I (Bovine) | |
25000 Units | |
RUO | |
DNASE1 | |
≥2,000 Kunitz units per mg dry weight. One Unit causes an increase in absorbance at 260nm of 0.001 per minute per ml at 25°C when acting upon highly polymerized DNA at pH 5.0. | |
None | |
Lyophilized |
Enzyme Activity | |
31.3kDa | |
Native protein chromatographically purified from Bovine Pancreas | |
Store at 4°C on dry atmosphere. | |
DNASE1 | |
Deoxyribonuclease I Assay; Nick translation; DNA mapping; Isolation of nuclear RNA and protein; Plasmid construction; RNA polymerase synthesis of RNA probes and RT-PCR. | |
Bovine | |
Native protein chromatographically purified from Bovine Pancreas |