Läs mer
Deoxysotalol, TRC
Produktkod. 30367490
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367490 10 mg
30355874 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30367490 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD24555010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 16974-42-8
Molekylformel C12H20N2O2S
Molekylvikt (g/mol) 256.36
Synonym Sotalol Impurity A
IUPAC-namn N-(4-(2-(isopropylamino)ethyl)phenyl)methanesulfonamide
LEDER O=S(NC1=CC=C(CCNC(C)C)C=C1)(C)=O
Specifikationer
Kemiskt namn eller material Deoxysotalol
Kvantitet 10 mg
InChI formel InChI=1S/C12H20N2O2S/c1-10(2)13-9-8-11-4-6-12(7-5-11)14-17(3,15)16/h4-7,10,13-14H,8-9H2,1-3H3
Formel vikt 256.12