Learn More
Abnova™ DEPDC1B Recombinant Protein
Brand:  Abnova™ H00055789-P01.10ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16131803
  • 4515.00 SEK / 10µg
Estimated Shipment: 23-05-2024
to see stock.
Description
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
AAH10904
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
46.31
8
Glutathione Sepharose 4 Fast Flow
10 ug
MPPLCDGFGTRTLMVQTFSRCILCSKDEVDLDELLAARLVTFLMDNYQEILKVPLALQTSIEERVAHLRRVQVKYPGADMDITLSAPSFCRQISPEEFEYQRSYGSQEPLAALLEEVITDAKLSNKEKKKKLKQFQKSYPEVYQERFPTPESAALLFPEKPKPKPQLLMWALKKPFQPFQRTRSFRM
BRCC3/FLJ11252/XTP1
DEPDC1B
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
55789
DEPDC1B (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DEPDC1B
Human
Recombinant
Solution