missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEPDC7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26517-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
DEPDC7 gene ontology annotations related to this gene include intracellular signal transduction; regulation of small GTPase mediated signal transduction.Specifications
DEPDC7 | |
Polyclonal | |
Unconjugated | |
DEPDC7 | |
DEP domain containing 7, DEPDC7, Protein TR2/D15, TR2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
91614 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96QD5 | |
DEPDC7 | |
DEPDC7 Fusion Protein Ag24094 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |