Läs mer
Des(1-cyclohexanol) Venlafaxine-d6, TRC
Produktkod. 30365172
Change view
Kvantitet:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30365172 2.5 mg
30360751 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30365172 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2885922.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1330046-00-8
Molekylformel C11H11D6NO
Molekylvikt (g/mol) 185.3
Synonym 4-Methoxy-N,N-(dimethyl-d6)benzeneethanamine, p-Methoxy-N,N-(dimethyl-d6) phenethylamine, N,N-(Dimethyl-d6)-2-(p-anisyl)ethylamine, N,N-(Dimethyl-d6)-4-methoxyphenethylamine
LEDER COC1=CC=C(CCN(C([2H])([2H])[2H])C([2H])([2H])[2H])C=C1
Specifikationer
Kemiskt namn eller material Des(1-cyclohexanol) Venlafaxine-d6
Kvantitet 2.5 mg
InChI formel InChI=1S/C11H17NO/c1-12(2)9-8-10-4-6-11(13-3)7-5-10/h4-7H,8-9H2,1-3H3/i1D3,2D3