Läs mer
Des(1-cyclohexanol) Venlafaxine, TRC
Produktkod. 30348878
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30348878 250 mg
1 options
Produktkod. 30348878 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD288590250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Des(1-cyclohexanol) Venlafaxine
CAS 775-33-7
Molekylformel C11 H17 N O
Kvantitet 250 mg
Synonym Benzeneethanamine, 4-methoxy-N,N-dimethyl- (9CI), Phenethylamine, p-methoxy-N,N-dimethyl- (7CI,8CI), 4-Methoxy-N,N-dimethylphenethylamine, N,N-Dimethyl-2-(p-anisyl)ethylamine, N,N-Dimethyl-4-methoxyphenethylamine, p-Methoxy-N,N-dimethylphenethylamine, 2-(4-Methoxyphenyl)-N,N-dimethylethanamine, Des(1-cyclohexanol) Venlafaxine, Venlafaxine Hydrochloride Imp. A (EP)
InChI formel InChI=1S/C11H17NO/c1-12(2)9-8-10-4-6-11(13-3)7-5-10/h4-7H,8-9H2,1-3H3
LEDER COc1ccc(CCN(C)C)cc1
IUPAC-namn 2-(4-methoxyphenyl)-N,N-dimethylethanamine
Molekylvikt (g/mol) 179.26
Formel vikt 179.131