Läs mer
Des-4,6-dimethoxypyrimidine Formimidamide Nicosulfuron, TRC
Produktkod. 30132965
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132965 50 mg
30135282 100 mg
30003908 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132965 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN39950550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2307738-55-0
Molekylformel C10 H14 N6 O4 S
Molekylvikt (g/mol) 314.321
Synonym Nicosulfuron Metabolite AUSN, 3-Pyridinecarboxamide, 2-[[[[(aminoiminomethyl)amino]carbonyl]amino]sulfonyl]-N,N-dimethyl-, IN-HYY21, Nicosulfuron Metabolite AUSN, 3-Pyridinecarboxamide, 2-[[[[(aminoiminomethyl)amino]carbonyl]amino]sulfonyl]-N,N-dimethyl-, 2-[[[[(Aminoiminomethyl)amino]carbonyl]amino]sulfonyl]-N,N-dimethyl-3-Pyridinecarboxamide
IUPAC-namn 2-(carbamimidoylcarbamoylsulfamoyl)-N,N-dimethylpyridine-3-carboxamide
LEDER CN(C)C(=O)c1cccnc1S(=O)(=O)NC(=O)NC(=N)N
Specifikationer
Kemiskt namn eller material Des-4,6-dimethoxypyrimidine Formimidamide Nicosulfuron
Kvantitet 50 mg
InChI formel InChI=1S/C10H14N6O4S/c1-16(2)8(17)6-4-3-5-13-7(6)21(19,20)15-10(18)14-9(11)12/h3-5H,1-2H3,(H5,11,12,14,15,18)
Formel vikt 314.0797