Läs mer
Des-dimethylpyrrolidine Eletriptan, TRC
Produktkod. 30356812
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30356812 100 mg
1 options
Produktkod. 30356812 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD289623100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Des-dimethylpyrrolidine Eletriptan
CAS 1225327-16-1
Molekylformel C16 H15 N O2 S
Kvantitet 100 mg
Synonym Des-dimethylpyrrolidine Eletriptan, 5-[2-(Phenylsulfonyl)ethyl]-1H-indole
InChI formel InChI=1S/C16H15NO2S/c18-20(19,15-4-2-1-3-5-15)11-9-13-6-7-16-14(12-13)8-10-17-16/h1-8,10,12,17H,9,11H2
LEDER O=S(=O)(CCc1ccc2[nH]ccc2c1)c3ccccc3
IUPAC-namn 5-[2-(benzenesulfonyl)ethyl]-1H-indole
Molekylvikt (g/mol) 285.361
Formel vikt 285.082