Läs mer
Des-glutamic Acid-(S)-Folinic Acid, TRC
Produktkod. 30315887
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30315887 10 mg
1 options
Produktkod. 30315887 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA15584010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (6S)-5-Formyltetrahydropteroic Acid
CAS 944737-05-7
Molekylformel C15 H16 N6 O4
Kvantitet 10 mg
Synonym 4-[[[(6S)-2-Amino-5-formyl-3,4,5,6,7,8-hexahydro-4-oxo-6-pteridinyl]methyl]amino]benzoic Acid, Benzoic acid, 4-[[[(6S)-2-amino-5-formyl-3,4,5,6,7,8-hexahydro-4-oxo-6-pteridinyl]methyl]amino]-, 4-[[[(6S)-2-Amino-5-formyl-3,4,5,6,7,8-hexahydro-4-oxo-6-pteridinyl]methyl]amino]benzoic acid, (6S)-5-Formyl-5,6,7,8-tetrahydropteroic acid, (6S)-5-Formyltet
InChI formel InChI=1S/C15H16N6O4/c16-15-19-12-11(13(23)20-15)21(7-22)10(6-18-12)5-17-9-3-1-8(2-4-9)14(24)25/h1-4,7,10,17H,5-6H2,(H,24,25)(H4,16,18,19,20,23)/t10-/m0/s1
LEDER NC1=NC(=O)C2=C(NC[C@H](CNc3ccc(cc3)C(=O)O)N2C=O)N1
IUPAC-namn 4-[[(6S)-2-amino-5-formyl-4-oxo-1,6,7,8-tetrahydropteridin-6-yl]methylamino]benzoic acid
Molekylvikt (g/mol) 344.325
Formel vikt 344.123