Läs mer
Des(isopropoxyethyl) Bisoprolol, TRC
Produktkod. 30134205
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134205 100 mg
30133886 25 mg
17998014 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134205 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD290125100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 62572-93-4
Molekylformel C13 H21 N O3
Molekylvikt (g/mol) 239.31
Synonym (RS)-1-(4-Hydroxymethylphenoxy)-3-isopropylaminopropan-2-ol, Bisoprolol Fumarate Imp. A (EP)
IUPAC-namn 1-[4-(hydroxymethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol
LEDER CC(C)NCC(O)COc1ccc(CO)cc1
Specifikationer
Kemiskt namn eller material (RS)-1-(4-Hydroxymethylphenoxy)-3-isopropylaminopropan-2-ol
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C13H21NO3/c1-10(2)14-7-12(16)9-17-13-5-3-11(8-15)4-6-13/h3-6,10,12,14-16H,7-9H2,1-2H3
Formel vikt 239.1521
Procent renhet >95%