Läs mer
Des-Methylcarbonate Hydroxy Flometoquin, TRC
Produktkod. 30362929
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30362929 50 mg
1 options
Produktkod. 30362929 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD11882550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Des-Methylcarbonate Hydroxy Flometoquin
CAS 1204518-38-6
Molekylformel C20H18F3NO3
Kvantitet 50 mg
Synonym 2-Ethyl-3,7-dimethyl-6-[4-(trifluoromethoxy)phenoxy]-4(1H)-quinolinone
InChI formel InChI=1S/C20H18F3NO3/c1-4-16-12(3)19(25)15-10-18(11(2)9-17(15)24-16)26-13-5-7-14(8-6-13)27-20(21,22)23/h5-10H,4H2,1-3H3,(H,24,25)
LEDER CCc1nc2cc(C)c(Oc3ccc(OC(F)(F)F)cc3)cc2c(O)c1C
IUPAC-namn 2-ethyl-3,7-dimethyl-6-[4-(trifluoromethoxy)phenoxy]-1H-quinolin-4-one
Molekylvikt (g/mol) 377.36
Formel vikt 377.1239