Läs mer
Des-methylformate Eplerenone, TRC
Produktkod. 30352292
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30352292 25 mg -
30365276 2.5 mg 2.5mg
30356282 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30352292 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29395525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 50394-27-9
Molekylformel C22H28O4
Molekylvikt (g/mol) 356.46
Synonym 9,11alpha-Epoxy-17-hydroxy-3-oxo-17alpha-gamma-lactone-pr50394-27-9egn-4-ene-21-carboxylic Acid, SC 12590
LEDER C[C@]12C[C@H]3O[C@@]34[C@@H](CCC5=CC(=O)CC[C@]45C)[C@@H]1CC[C@@]26CCC(=O)O6
Specifikationer
Kemiskt namn eller material Des-methylformate Eplerenone
Kvantitet 25 mg
InChI formel InChI=1S/C22H28O4/c1-19-8-5-14(23)11-13(19)3-4-16-15-6-9-21(10-7-18(24)26-21)20(15,2)12-17-22(16,19)25-17/h11,15-17H,3-10,12H2,1-2H3
Formel vikt 356.1988