Läs mer
Des(methylpiperazinyl-N-methyl) Imatinib Dimer Impurity, TRC
Produktkod. 30354145
Change view
Kvantitet:
2.5 mg
Produktkod. Kvantitet
30354145 2.5 mg
1 options
Produktkod. 30354145 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2918902.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4,4'-[1,4-Piperazinediylbis(methylene)]bis[N-[4-methyl-3-[(4-(3-pyridinyl)-2-pyrimidinyl)amino]phenyl]benzamide]
CAS 1365802-18-1
Molekylformel C52 H48 N12 O2
Kvantitet 2.5 mg
Synonym Benzamide, 4,4'-[1,4-piperazinediylbis(methylene)]bis[N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]-, 4,4'-[1,4-Piperazinediylbis(methylene)]bis[N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]benzamide], Des(methylpiperazinyl-N-methyl) Imatinib Dimer Impurity
InChI formel InChI=1S/C52H48N12O2/c1-35-7-17-43(29-47(35)61-51-55-23-19-45(59-51)41-5-3-21-53-31-41)57-49(65)39-13-9-37(10-14-39)33-63-25-27-64(28-26-63)34-38-11-15-40(16-12-38)50(66)58-44-18-8-36(2)48(30-44)62-52-56-24-20-46(60-52)42-6-4-22-54-32-42/h3-24,29-32H,25-28,33-34H2,1-2H3,(H,57,65)(H,58,66)(H,55,59,61)(H,56,60,62)
LEDER Cc1ccc(NC(=O)c2ccc(CN3CCN(Cc4ccc(cc4)C(=O)Nc5ccc(C)c(Nc6nccc(n6)c7cccnc7)c5)CC3)cc2)cc1Nc8nccc(n8)c9cccnc9
IUPAC-namn N-[4-methyl-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]-4-[[4-[[4-[[4-methyl-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]carbamoyl]phenyl]methyl]piperazin-1-yl]methyl]benzamide
Molekylvikt (g/mol) 873.02
Formel vikt 872.4023