Läs mer
Des-(N-2-pyridyl-β-alanine Ethyl Ester) Dabigatran ethyl ester hydrochloride, TRC
Produktkod. 30322916
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30322916 50 mg
1 options
Produktkod. 30322916 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA61137050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(((4-(Aminoiminomethyl)phenyl)amino)methyl)-1-methyl-1H-benzimidazole-5-carboxylic Acid Ethyl Ester Hydrochloride
CAS 1422435-39-9
Molekylformel C19H21N5O2 . (HCl)
Kvantitet 50 mg
Synonym 2-[[[4-(Aminoiminomethyl)phenyl]amino]methyl]-1-methyl-1H-benzimidazole-5-carboxylic Acid Ethyl Ester Hydrochloride
InChI formel InChI=1S/C19H21N5O2.ClH/c1-3-26-19(25)13-6-9-16-15(10-13)23-17(24(16)2)11-22-14-7-4-12(5-8-14)18(20)21;/h4-10,22H,3,11H2,1-2H3,(H3,20,21);1H
LEDER Cl.CCOC(=O)c1ccc2c(c1)nc(CNc3ccc(cc3)C(=N)N)n2C
IUPAC-namn ethyl 2-[(4-carbamimidoylanilino)methyl]-1-methylbenzimidazole-5-carboxylate;hydrochloride
Molekylvikt (g/mol) 387.9
Formel vikt 387.1462