Läs mer
Des-(N-methyl-5-tetrazolethiolyl)furolactone Cefoperazone, TRC
Produktkod. 30005867
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30005867 10 mg
1 options
Produktkod. 30005867 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29432510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Renhet >95% (HPLC)
CAS 73240-08-1
Formatera Neat
Molekylformel C23H23N5O8S
Molekylvikt (g/mol) 529.52
InChI-nyckel RFNBMKDSMYYHTH-VTJXTGGHSA-N
LEDER [H][C@]12SCC3=C(N1C(=O)[C@H]2NC(=O)[C@H](NC(=O)N1CCN(CC)C(=O)C1=O)C1=CC=C(O)C=C1)C(=O)OC3
IUPAC-namn (2R)-N-[(4R,5R)-3,11-dioxo-10-oxa-6-thia-2-azatricyclo[6.3.0.0²,⁵]undec-1(8)-en-4-yl]-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-(4-hydroxyphenyl)acetamide
Produkttyp Impurity
Kvantitet 10 mg
Frakt skick Room Temperature
Synonym [5aR-[5aα,6β(R*)]]-4-Ethyl-N-[1-(4-hydroxyphenyl)-2-oxo-2-[(1,4,5a,6-tetrahydro-