Läs mer
Des(N,N-dimethylethanamine) Orphenadrine Dimer, TRC
Produktkod. 30358942
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30358942 50 mg
1 options
Produktkod. 30358942 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29333550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Des(N,N-dimethylethanamine) Orphenadrine Dimer
CAS 40673-52-7
Molekylformel C28 H26 O
Kvantitet 50 mg
Synonym Bis(o-methyl-α-phenylbenzyl) ether, Bis(phenyl-o-tolylmethyl) ether
InChI formel InChI=1S/C28H26O/c1-21-13-9-11-19-25(21)27(23-15-5-3-6-16-23)29-28(24-17-7-4-8-18-24)26-20-12-10-14-22(26)2/h3-20,27-28H,1-2H3
LEDER O(C(C=1C=CC=CC1)C=2C=CC=CC2C)C(C=3C=CC=CC3)C=4C=CC=CC4C
IUPAC-namn 1-methyl-2-[[(2-methylphenyl)-phenylmethoxy]-phenylmethyl]benzene
Molekylvikt (g/mol) 378.5
Formel vikt 378.198