Läs mer
Desacetyl Desformylvincristine, TRC
Produktkod. 30367264
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30367264 5 mg
1 options
Produktkod. 30367264 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2967555MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Desacetyl Desformylvincristine
CAS 55324-83-9
Molekylformel C43H54N4O8
Kvantitet 5 mg
Synonym O4-Deacetyl-1-demethylvincaleukoblastine
InChI formel InChI=1S/C43H54N4O8/c1-6-39(51)21-25-22-42(37(49)54-4,33-27(13-17-46(23-25)24-39)26-11-8-9-12-30(26)44-33)29-19-28-31(20-32(29)53-3)45-34-41(28)15-18-47-16-10-14-40(7-2,35(41)47)36(48)43(34,52)38(50)55-5/h8-12,14,19-20,25,34-36,44-45,48,51-52H,6-7,13,15-18,21-24H2,1-5H3
LEDER CC[C@]1(O)C[C@@H]2CN(CCc3c([nH]c4ccccc34)[C@@](C2)(C(=O)OC)c5cc6c(N[C@H]7[C@](O)([C@H](O)[C@]8(CC)C=CCN9CC[C@]67C89)C(=O)OC)cc5OC)C1
Molekylvikt (g/mol) 754.91