Läs mer
Desacetyl-N,O-descarbonyl Linezolid (Technical Grade), TRC
Produktkod. 30356300
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30356300 25 mg
1 options
Produktkod. 30356300 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28828525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Desacetyl-N,O-descarbonyl Linezolid
CAS 333753-72-3
Molekylformel C13H20FN3O2
Kvantitet 25 mg
Synonym (2S)-1-Amino-3-[[3-fluoro-4-(4-morpholinyl)phenyl]amino]-2-propanol
InChI formel InChI=1S/C13H20FN3O2/c14-12-7-10(16-9-11(18)8-15)1-2-13(12)17-3-5-19-6-4-17/h1-2,7,11,16,18H,3-6,8-9,15H2
LEDER NC[C@H](O)CNc1ccc(N2CCOCC2)c(F)c1
Molekylvikt (g/mol) 269.32
Formel vikt 269.154