Läs mer
Desacetylcinobufagin, MedChemExpress
Produktkod. 16471568
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
Förpackningsstorlek:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16471568 5 mg 5mg
16469158 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16471568 Leverantör Medchem Express Leverantörsnummer HYN0881
MedChemExpress Desacetylcinobufagin is a natural compound used for microbial transformation.
Desacetylcinobufagin is a natural compound used for microbial transformation.
Specifikationer
Kemiskt namn eller material Desacetylcinobufagin
CAS 4026-95-3
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C24H32O5
Kvantitet 5 mg
Synonym Deacetylcinobufagin
Löslighetsinformation DMSO : ≥ 26 mg/mL (64.92 mM)
LEDER C[C@]([C@@H](C(C=C1)=COC1=O)[C@H]2O)(CC[C@@]3([H])[C@@]4([H])CC[C@@]5([H])[C@@]3(CC[C@H](O)C5)C)[C@@]64[C@@H]2O6
Molekylvikt (g/mol) 400.51
Formel vikt 400.51
Procent renhet 99.04%
Kvalitet Research
Fysisk form Solid