Läs mer
Desaminosulfonyl Dorzolamide Hydrochloride, TRC
Produktkod. 30352690
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30352690 10 mg
1 options
Produktkod. 30352690 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28895510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Desaminosulfonyl Dorzolamide Hydrochloride
CAS 897657-77-1
Molekylformel C10H15NO2S2 . HCl
Kvantitet 10 mg
Synonym (4S,6S)-N-Ethyl-5,6-dihydro-6-methyl-4H-thieno[2,3-b]thiopyran-4-amine 7,7-dioxide hydrochloride
InChI formel InChI=1S/C10H15NO2S2.ClH/c1-3-11-9-6-7(2)15(12,13)10-8(9)4-5-14-10;/h4-5,7,9,11H,3,6H2,1-2H3;1H/t7-,9-;/m0./s1
LEDER Cl.CCN[C@H]1C[C@H](C)S(=O)(=O)c2sccc12
Molekylvikt (g/mol) 281.82
Formel vikt 281.0311