Läs mer
Descarboxyl Levofloxacin, TRC
Produktkod. 30004721
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004721 25 mg
30011446 50 mg
17992095 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30004721 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28848025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 178964-53-9
Molekylformel C17 H20 F N3 O2
Molekylvikt (g/mol) 317.36
Synonym (3S)-9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7H-pyrido[1,2,3-de]-1,4-benzoxazin-7-one
IUPAC-namn (2S)-7-fluoro-2-methyl-6-(4-methylpiperazin-1-yl)-4-oxa-1-azatricyclo[7.3.1.05,13]trideca-5(13),6,8,11-tetraen-10-one
LEDER C[C@H]1COc2c(N3CCN(C)CC3)c(F)cc4C(=O)C=CN1c24
Specifikationer
Kemiskt namn eller material Decarboxyl levofloxacin
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C17H20FN3O2/c1-11-10-23-17-15-12(14(22)3-4-21(11)15)9-13(18)16(17)20-7-5-19(2)6-8-20/h3-4,9,11H,5-8,10H2,1-2H3/t11-/m0/s1
Formel vikt 317.154
Procent renhet >95%