Läs mer
Deschloro-1,2-dihydro-2-oxo Clomiphene Hydrochloride Salt, TRC
Produktkod. 30003713
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30003713 50 mg
30009463 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30003713 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28904050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5635-70-1
Molekylformel C26 H29 N O2 . Cl H
Molekylvikt (g/mol) 423.97
Synonym 2-[p-[2-(diethylamino)ethoxy]phenyl]-2-phenylacetophenone Hydrochloride (7CI,8CI), (2RS)-2-[4-[2-(diethylamino)ethoxy]phenyl]-1,2-diphenylethanone Hydrochloride, Clomifene Citrate Imp. C (EP) as Hydrochloride
IUPAC-namn 2-[4-[2-(diethylamino)ethoxy]phenyl]-1,2-diphenylethanone;hydrochloride
LEDER Cl.CCN(CC)CCOc1ccc(cc1)C(C(=O)c2ccccc2)c3ccccc3
Specifikationer
Kemiskt namn eller material Deschloro-1,2-dihydro-2-oxo Clomiphene hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C26H29NO2.ClH/c1-3-27(4-2)19-20-29-24-17-15-22(16-18-24)25(21-11-7-5-8-12-21)26(28)23-13-9-6-10-14-23;/h5-18,25H,3-4,19-20H2,1-2H3;1H
Formel vikt 423.1965
Procent renhet >95%