Läs mer
Deschloro Clomiphene (E/Z Mixture), TRC
Produktkod. 30365553
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30365553 100 mg
1 options
Produktkod. 30365553 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD288980100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Deschloro Clomiphene (E/Z Mixture)
CAS 19957-52-9
Molekylformel C26H29NO
Kvantitet 100 mg
Synonym 2-[4-(1,2-Diphenylethenyl)phenoxy]-N,N-diethyl-ethanamine, 2-[p-(1,2-Diphenylvinyl)phenoxy]-triethylamine, E.I.P.W. 103, α-[4-(2-Diethylaminoethoxy)phenyl]stilbene
InChI formel InChI=1S/C26H29NO/c1-3-27(4-2)19-20-28-25-17-15-24(16-18-25)26(23-13-9-6-10-14-23)21-22-11-7-5-8-12-22/h5-18,21H,3-4,19-20H2,1-2H3/b26-21+
LEDER CCN(CC)CCOC1=CC=C(C=C1)C(=CC2=CC=CC=C2)C3=CC=CC=C3
Molekylvikt (g/mol) 371.51
Formel vikt 371.2249