Läs mer
Deschloro Clomiphene (E/Z Mixture), TRC
Produktkod. 30363437
Change view
Kvantitet:
25 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30363437 500 mg
30361001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30363437 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD288980500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Deschloro Clomiphene (E/Z Mixture)
CAS 19957-52-9
Molekylformel C26H29NO
Kvantitet 500 mg
Synonym 2-[4-(1,2-Diphenylethenyl)phenoxy]-N,N-diethyl-ethanamine, 2-[p-(1,2-Diphenylvinyl)phenoxy]-triethylamine, E.I.P.W. 103, α-[4-(2-Diethylaminoethoxy)phenyl]stilbene
InChI formel InChI=1S/C26H29NO/c1-3-27(4-2)19-20-28-25-17-15-24(16-18-25)26(23-13-9-6-10-14-23)21-22-11-7-5-8-12-22/h5-18,21H,3-4,19-20H2,1-2H3/b26-21+
LEDER CCN(CC)CCOC1=CC=C(C=C1)C(=CC2=CC=CC=C2)C3=CC=CC=C3
Molekylvikt (g/mol) 371.51
Formel vikt 371.2249