Läs mer
Descyano Cimetidine Dihydrochloride Salt, TRC
Produktkod. 30366799
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30366799 250 mg
1 options
Produktkod. 30366799 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD290645250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Methyl-3-[2-([(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl)ethyl]guanidine Dihydrochloride
CAS 59660-24-1
Anteckningar om renhetsgrad HPLC
Molekylformel C9 H17 N5 S . 2 Cl H
Kvantitet 250 mg
Synonym 1-Methyl-3-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]guanidine Dihydrochloride, Cimetidine Hydrochloride Imp. D (EP) as Dihydrochloride
InChI formel InChI=1S/C9H17N5S.2ClH/c1-7-8(14-6-13-7)5-15-4-3-12-9(10)11-2;;/h6H,3-5H2,1-2H3,(H,13,14)(H3,10,11,12);2*1H
LEDER Cl.Cl.CNC(=N)NCCSCc1nc[nH]c1C
IUPAC-namn 1-methyl-3-[2-[(5-methyl-1H-imidazol-4-yl)methylsulfanyl]ethyl]guanidine;dihydrochloride
Molekylvikt (g/mol) 300.25
Formel vikt 299.0738