Läs mer
Descyano Cimetidine Dihydrochloride Salt, TRC
Produktkod. 30362363
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30362363 25 mg
30349849 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30362363 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29064525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Methyl-3-[2-([(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl)ethyl]guanidine Dihydrochloride
CAS 59660-24-1
Anteckningar om renhetsgrad HPLC
Molekylformel C9 H17 N5 S . 2 Cl H
Kvantitet 25 mg
Synonym 1-Methyl-3-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]guanidine Dihydrochloride, Cimetidine Hydrochloride Imp. D (EP) as Dihydrochloride
InChI formel InChI=1S/C9H17N5S.2ClH/c1-7-8(14-6-13-7)5-15-4-3-12-9(10)11-2;;/h6H,3-5H2,1-2H3,(H,13,14)(H3,10,11,12);2*1H
LEDER Cl.Cl.CNC(=N)NCCSCc1nc[nH]c1C
IUPAC-namn 1-methyl-3-[2-[(5-methyl-1H-imidazol-4-yl)methylsulfanyl]ethyl]guanidine;dihydrochloride
Molekylvikt (g/mol) 300.25
Formel vikt 299.0738