Läs mer
Descyclopropyl Abacavir, TRC
Produktkod. 30367963
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30367963 10 mg
1 options
Produktkod. 30367963 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28886510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Descyclopropyl Abacavir
CAS 124752-25-6
Molekylformel C11 H14 N6 O
Kvantitet 10 mg
Synonym 2-Cyclopentene-1-methanol, 4-(2,6-diamino-9H-purin-9-yl)-, (1S,4R)-, 2-Cyclopentene-1-methanol, 4-(2,6-diamino-9H-purin-9-yl)-, (1S-cis)-, Descyclopropyl Abacavir, Abacavir Related Compound A (USP), Abacavir Sulfate Imp. C (Ph. Int.), Abacavir Sulfate Imp C (EP)
InChI formel InChI=1S/C11H14N6O/c12-9-8-10(16-11(13)15-9)17(5-14-8)7-2-1-6(3-7)4-18/h1-2,5-7,18H,3-4H2,(H4,12,13,15,16)/t6-,7+/m1/s1
LEDER Nc1nc(N)c2ncn([C@@H]3C[C@H](CO)C=C3)c2n1
IUPAC-namn [(1S,4R)-4-(2,6-diaminopurin-9-yl)cyclopent-2-en-1-yl]methanol
Molekylvikt (g/mol) 246.27
Formel vikt 246.1229