Läs mer
Descyclopropyl Lenvatinib, TRC
Produktkod. 30350726
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30350726 50 mg
1 options
Produktkod. 30350726 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28815050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Descyclopropyl Lenvatinib
CAS 417719-51-8
Molekylformel C18H15ClN4O4
Kvantitet 50 mg
Synonym 4-[4-[(Aminocarbonyl)amino]-3-chlorophenoxy]-7-methoxy-6-quinolinecarboxamide, Lenvatinib M1
InChI formel InChI=1S/C18H15ClN4O4/c1-26-16-8-14-10(7-11(16)17(20)24)15(4-5-22-14)27-9-2-3-13(12(19)6-9)23-18(21)25/h2-8H,1H3,(H2,20,24)(H3,21,23,25)
LEDER COc1cc2nccc(Oc3ccc(NC(=O)N)c(Cl)c3)c2cc1C(=O)N
Molekylvikt (g/mol) 386.79