Läs mer
Desethyl Azilsartan Medoxomil, TRC
Produktkod. 30349829
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30349829 100 mg
30365433 50 mg
30363249 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30349829 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD289258100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1417576-00-1
Molekylformel C28H20N4O8
Molekylvikt (g/mol) 540.48
Synonym 3-[[2'-(2,5-Dihydro-5-oxo-1,2,4-oxadiazol-3-yl)[1,1'-biphenyl]-4-yl]methyl]-2,3-dihydro-2-oxo-1H-benzimidazole-4-carboxylic Acid (5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl Ester
LEDER CC1=C(COC(=O)c2cccc3NC(=O)N(Cc4ccc(cc4)c5ccccc5C6=NOC(=O)N6)c23)OC(=O)O1
Specifikationer
Kemiskt namn eller material Desethyl Azilsartan Medoxomil
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C28H20N4O8/c1-15-22(39-28(36)38-15)14-37-25(33)20-7-4-8-21-23(20)32(26(34)29-21)13-16-9-11-17(12-10-16)18-5-2-3-6-19(18)24-30-27(35)40-31-24/h2-12H,13-14H2,1H3,(H,29,34)(H,30,31,35)
Formel vikt 540.1281
Procent renhet >95%