Läs mer
Desethyl Chloroquine, TRC
Produktkod. 17903375
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17903375 25 mg -
17982565 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17903375 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28873525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1476-52-4
Molekylformel C16 H22 Cl N3
Molekylvikt (g/mol) 291.82
Synonym N4-(7-Chloro-4-quinolinyl)-N1-ethyl-1,4-pentanediamine, Deethylchloroquine, Monodeethylchloroquine, NSC 13254, WR 29623, Desethyl Cloroquine
IUPAC-namn 4-N-(7-chloroquinolin-4-yl)-1-N-ethylpentane-1,4-diamine
LEDER CCNCCCC(C)Nc1ccnc2cc(Cl)ccc12
Specifikationer
Kemiskt namn eller material Desethylchloroquine
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C16H22ClN3/c1-3-18-9-4-5-12(2)20-15-8-10-19-16-11-13(17)6-7-14(15)16/h6-8,10-12,18H,3-5,9H2,1-2H3,(H,19,20)
Formel vikt 291.1502
Procent renhet >95%