Läs mer
Desethylcarbamoyl Cabergoline, TRC
Produktkod. 30358685
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30358685 50 mg
30360714 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30358685 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28886050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 85329-86-8
Molekylformel C23 H32 N4 O
Molekylvikt (g/mol) 380.53
Synonym (8β)-N-[3-(dimethylamino)propyl]-6-(2-propen-1-yl)ergoline-8-carboxamide, FCE 21590, (6aR,9R,10aR)-N-[3-(Dimethylamino)propyl]-7-(prop-2-enyl)-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinoline-9-carboxamide, Cabergoline Imp. D (EP)
IUPAC-namn (6aR,9R,10aR)-N-[3-(dimethylamino)propyl]-7-prop-2-enyl-6,6a,8,9,10,10a-hexahydro-4H-indolo[4,3-fg]quinoline-9-carboxamide
LEDER CN(C)CCCNC(=O)[C@@H]1C[C@H]2[C@@H](Cc3c[nH]c4cccc2c34)N(CC=C)C1
Specifikationer
Kemiskt namn eller material Desethylcarbamoyl Cabergoline
Kvantitet 50 mg
InChI formel InChI=1S/C23H32N4O/c1-4-10-27-15-17(23(28)24-9-6-11-26(2)3)12-19-18-7-5-8-20-22(18)16(14-25-20)13-21(19)27/h4-5,7-8,14,17,19,21,25H,1,6,9-13,15H2,2-3H3,(H,24,28)/t17-,19-,21-/m1/s1
Formel vikt 380.2576