Läs mer
Desethylclomifene Hydrochloride, Mikromol™
Produktkod. 30051285
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051285 100 mg
30050682 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051285 Leverantör Mikromol Leverantörsnummer MM0650.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Desethylclomifene Hydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Desethylclomifene Hydrochloride
CAS 1310815-19-0
Analyt- eller komponentnamn Desethylclomifene Hydrochloride
Frakt skick Room Temperature
Molekylformel C24 H24 Cl N O . Cl H
Certifikat och efterlevnad ISO 17025
Synonym Desethylclomifene Hydrochloride
InChI formel InChI=1S/C24H24ClNO.ClH/c1-2-26-17-18-27-22-15-13-20(14-16-22)23(19-9-5-3-6-10-19)24(25)21-11-7-4-8-12-21;/h3-16,26H,2,17-18H2,1H3;1H
LEDER Cl.CCNCCOc1ccc(cc1)C(=C(Cl)c2ccccc2)c3ccccc3
IUPAC-namn 2-[4-(2-chloro-1,2-diphenylethenyl)phenoxy]-N-ethylethanamine;hydrochloride
Molekylvikt (g/mol) 414.37
Kvantitet 100 mg
Formel vikt 413.1313 g/mol