Läs mer
Desethyloxybutynin Hydrochloride, Mikromol™
Produktkod. 17982036
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17982036 100 mg
17942296 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17982036 Leverantör Mikromol Leverantörsnummer MM0047.11
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Desethyloxybutynin Hydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Desethyloxybutynin Hydrochloride
CAS 81039-77-2
Analyt- eller komponentnamn Desethyloxybutynin Hydrochloride
Frakt skick Room Temperature
Molekylformel C20 H27 N O3 . Cl H
Certifikat och efterlevnad ISO 17025
Synonym Desethyloxybutynin Hydrochloride
InChI formel InChI=1S/C20H27NO3.ClH/c1-2-21-15-9-10-16-24-19(22)20(23,17-11-5-3-6-12-17)18-13-7-4-8-14-18;/h3,5-6,11-12,18,21,23H,2,4,7-8,13-16H2,1H3;1H
LEDER Cl.CCNCC#CCOC(=O)C(O)(C1CCCCC1)c2ccccc2
IUPAC-namn 4-(ethylamino)but-2-ynyl 2-cyclohexyl-2-hydroxy-2-phenylacetate;hydrochloride
Molekylvikt (g/mol) 365.89
Kvantitet 100 mg
Formel vikt 365.1758 g/mol