Läs mer
Desfluoro Fluvoxamine, TRC
Produktkod. 30358691
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30358691 10 mg
30352534 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30358691 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28997510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1217214-94-2
Molekylformel C15 H22 F2 N2 O2
Molekylvikt (g/mol) 300.34
Synonym 1-Pentanone, 1-[4-(difluoromethyl)phenyl]-5-methoxy-, O-(2-aminoethyl)oxime, 2-[[[1-[4-(Difluoromethyl)phenyl]-5-methoxypentylidene]amino]oxy]ethanamine, Desfluorofluvoxamine, Defluoro fluvoxamine
IUPAC-namn 2-[[1-[4-(difluoromethyl)phenyl]-5-methoxypentylidene]amino]oxyethanamine
LEDER COCCCCC(=NOCCN)c1ccc(cc1)C(F)F
Specifikationer
Kemiskt namn eller material desfluoro Fluvoxamine
Kvantitet 10 mg
InChI formel InChI=1S/C15H22F2N2O2/c1-20-10-3-2-4-14(19-21-11-9-18)12-5-7-13(8-6-12)15(16)17/h5-8,15H,2-4,9-11,18H2,1H3
Formel vikt 300.1649