Läs mer
Desfluoro Fluvoxketone, TRC
Produktkod. 30353966
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30353966 25 mg
30349651 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30353966 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28992525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391053-78-3
Molekylformel C13H16F2O2
Molekylvikt (g/mol) 242.26
Synonym (4-Methoxybutyl)(4-difluoromethylphenyl)methanone, Desfluoro EP Impurity D 5-Methoxy-1-[4-(difluoromethyl)phenyl]-1-pentanone
LEDER COCCCCC(=O)C1=CC=C(C=C1)C(F)F
Specifikationer
Kemiskt namn eller material Desfluoro Fluvoxketone
Kvantitet 25 mg
InChI formel InChI=1S/C13H16F2O2/c1-17-9-3-2-4-12(16)10-5-7-11(8-6-10)13(14)15/h5-8,13H,2-4,9H2,1H3
Formel vikt 242.1118