Läs mer
Desfluoroatorvastatin, TRC
Produktkod. 30379917
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30379917 1 mg
1 options
Produktkod. 30379917 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD7917501MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Desfluoroatorvastatin
CAS 433289-84-0
Molekylformel C33 H36 N2 O5
Kvantitet 1 mg
Synonym (3R,5R)-3,5-dihydroxy-7-[5-(1-methylethyl)-2,3-diphenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl]heptanoic acid (desfluoroatorvastatin), (betaR,deltaR)-beta,delta-dihydroxy-2-(1-methylethyl)-4,5-diphenyl-3-[(phenylamino)carbonyl]-1H-Pyrrole-1-heptanoic acid, Atorvastatin Calcium Trihydrate Imp. A (EP)
InChI formel InChI=1S/C33H36N2O5/c1-22(2)31-30(33(40)34-25-16-10-5-11-17-25)29(23-12-6-3-7-13-23)32(24-14-8-4-9-15-24)35(31)19-18-26(36)20-27(37)21-28(38)39/h3-17,22,26-27,36-37H,18-21H2,1-2H3,(H,34,40)(H,38,39)/t26-,27-/m1/s1
LEDER CC(C)c1c(C(=O)Nc2ccccc2)c(c3ccccc3)c(c4ccccc4)n1CC[C@@H](O)C[C@@H](O)CC(=O)O
IUPAC-namn (3R,5R)-7-[2,3-diphenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-3,5-dihydroxyheptanoic acid
Molekylvikt (g/mol) 540.65
Formel vikt 540.2624