Läs mer
Desfluoroenrofloxacin Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Desfluoroenrofloxacin Hydrochloride |
| CAS | 2732310-93-7 |
| Molekylformel | C19 H23 N3 O3 . Cl H |
| Kvantitet | 250 mg |
| Synonym | 1-Cyclopropyl-7-(4-ethylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid Hydrochloride, Enrofloxacin Imp. C (EP) as Hydrochloride, Desfluoroenrofloxacin Hydrochloride, 3-Quinolinecarboxylic acid, 1-cyclopropyl-7-(4-ethyl-1-piperazinyl)-1,4-dihydro-4-oxo-, hydrochloride (1:1) (ACI) |
| InChI formel | InChI=1S/C19H23N3O3.ClH/c1-2-20-7-9-21(10-8-20)14-5-6-15-17(11-14)22(13-3-4-13)12-16(18(15)23)19(24)25;/h5-6,11-13H,2-4,7-10H2,1H3,(H,24,25);1H |
| LEDER | Cl.CCN1CCN(CC1)c2ccc3C(=O)C(=CN(C4CC4)c3c2)C(=O)O |
| IUPAC-namn | 1-cyclopropyl-7-(4-ethylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrochloride |
| Molekylvikt (g/mol) | 377.87 |
| Formel vikt | 377.1506 |