Läs mer
Desfluoromethyl Fluticasone Propionate Disulfide, TRC
Produktkod. 17931675
Change view
Kvantitet:
1 mg
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17931675 2.5 mg 2.5mg
17923995 1 mg -
17903855 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17931675 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2898902.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 201812-64-8
Molekylformel C48 H58 F4 O10 S2
Molekylvikt (g/mol) 935.09
Synonym Androsta-1,4-dien-3-one, 17,17'-(dithiodicarbonyl)bis[6,9-difluoro-11-hydroxy-16-methyl-17-(1-oxopropoxy)-, (6α,11β,16α,17α)-(6'α,11'β,16'α,17'α)-, 17,17′-(disulfanediyldicarbonyl)bis(6α,9-difluoro-11β-hydroxy-16α-methyl-3-oxoandrosta-1,4-dien-17α-yl) dipropanoate, Fluticasone Propionate Imp. H (EP), unidentified impurity with RRT 1.23, fluticasone dipropionate disulphide (GR 68620X), disulphide dipropionate dithioketone (GR 247095X), dithio (GR 269949X)
IUPAC-namn [(6S,8S,9R,10S,11S,13S,14S,16R,17R)-17-[[(6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17-carbonyl]disulfanyl]carbonyl-6,9-difluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] propanoate
LEDER CCC(=O)O[C@@]1([C@H](C)C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)SSC(=O)[C@@]5(OC(=O)CC)[C@H](C)C[C@H]6[C@@H]7C[C@H](F)C8=CC(=O)C=C[C@]8(C)[C@@]7(F)[C@@H](O)C[C@]56C
Specifikationer
Kemiskt namn eller material Desfluoromethyl Fluticasone Propionate Disulfide
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C48H58F4O10S2/c1-9-37(57)61-47(23(3)15-27-29-19-33(49)31-17-25(53)11-13-41(31,5)45(29,51)35(55)21-43(27,47)7)39(59)63-64-40(60)48(62-38(58)10-2)24(4)16-28-30-20-34(50)32-18-26(54)12-14-42(32,6)46(30,52)36(56)22-44(28,48)8/h11-14,17-18,23-24,27-30,33-36,55-56H,9-10,15-16,19-22H2,1-8H3/t23-,24-,27+,28+,29+,30+,33+,34+,35+,36+,41+,42+,43+,44+,45+,46+,47+,48+/m1/s1
Formel vikt 934.3408
Procent renhet >95%