Läs mer
Tocris Bioscience™ Desformylflustrabromine hydrochloride
GREENER_CHOICE
Produktkod. 18035682
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18035682 10 mg
18045062 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18035682 Leverantör Tocris Bioscience™ Leverantörsnummer 3328/10
Positive allosteric modulator of α4β2; also muscle-type nAChR inhibitor
Positive allosteric modulator of nicotinic α4β2 receptors; selectively increases the ionic current through α4β2 in the presence of ACh. Displays 14.7-fold selectivity for α4β2 over homomeric (α7) receptors. Moderately cytotoxic in HCT-116 cells. Also inhibits human muscle (αβεδ) and Torpedo (αβγδ) nAChRs (IC50 values are 1.0 and 0.1 μM, respectively) by binding in the ion channel.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 951322-11-5
Molekylformel C16H22BrClN2
Specifikationer
Kemiskt namn eller material Desformylflustrabromine hydrochloride
Kvantitet 10 mg
Formel vikt 357.72
Renhet >98%
Molekylvikt (g/mol) 357.72