Läs mer
Deshydroxy Amino Ticagrelor Acetonide, TRC
Produktkod. 30356060
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30356060 100 mg
1 options
Produktkod. 30356060 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD192180100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Deshydroxy Amino Ticagrelor Acetonide
CAS 1902184-01-3
Molekylformel C26H33F2N7O3S
Kvantitet 100 mg
Synonym 3-((3aS,4R,6S,6aR)-6-(2-aminoethoxy)-2,2-dimethyltetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-yl)-N-((1R,2S)-2-(3,4-difluorophenyl)cyclopropyl)-5-(propylthio)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-7-amine
InChI formel InChI=1S/C26H33F2N7O3S/c1-4-9-39-25-31-23(30-17-11-14(17)13-5-6-15(27)16(28)10-13)20-24(32-25)35(34-33-20)18-12-19(36-8-7-29)22-21(18)37-26(2,3)38-22/h5-6,10,14,17-19,21-22H,4,7-9,11-12,29H2,1-3H3,(H,30,31,32)/t14-,17+,18+,19-,21-,22+/m0/s1
LEDER CCCSc1nc(N[C@@H]2C[C@H]2c3ccc(F)c(F)c3)c4nnn([C@@H]5C[C@H](OCCN)[C@H]6OC(C)(C)O[C@@H]56)c4n1
IUPAC-namn 3-[(3aR,4S,6R,6aS)-4-(2-aminoethoxy)-2,2-dimethyl-4,5,6,6a-tetrahydro-3aH-cyclopenta[d][1,3]dioxol-6-yl]-N-[(1R,2S)-2-(3,4-difluorophenyl)cyclopropyl]-5-propylsulfanyltriazolo[4,5-d]pyrimidin-7-amine
Molekylvikt (g/mol) 561.65
Formel vikt 561.2334