Läs mer
Deshydroxymethyl Losartan Trifluoroacetic Acid Salt, TRC
Produktkod. 30360179
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30360179 250 mg
1 options
Produktkod. 30360179 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD290020250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Deshydroxymethyl Losartan Trifluoroacetic Acid Salt
CAS 1215727-33-5
Molekylformel C21 H21 Cl N6 . C2 H F3 O2
Kvantitet 250 mg
Synonym Losartan Impurity C
InChI formel InChI=1S/C21H21ClN6.C2HF3O2/c1-2-3-8-20-23-19(22)14-28(20)13-15-9-11-16(12-10-15)17-6-4-5-7-18(17)21-24-26-27-25-21;3-2(4,5)1(6)7/h4-7,9-12,14H,2-3,8,13H2,1H3,(H,24,25,26,27);(H,6,7)
LEDER CCCCc1nc(Cl)cn1Cc2ccc(cc2)c3ccccc3c4nn[nH]n4.OC(=O)C(F)(F)F
IUPAC-namn 5-[2-[4-[(2-butyl-4-chloro-imidazol-1-yl)methyl]phenyl]phenyl]-2H-tetrazole;2,2,2-trifluoroacetic acid
Molekylvikt (g/mol) 506.91
Formel vikt 506.1445