Läs mer
DESI1 Polyclonal Antibody
DESI1 Polyclonal Antibody
Brand: Invitrogen PA571343
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
DESI1 Polyclonal Antibody for Western Blot
Specifikationer
| DESI1 | |
| Polyclonal | |
| Unconjugated | |
| DESI1 | |
| D15Wsu75e; DESI1; DeSI-1; DESI2; desumoylating isopeptidase 1; desumoylating isopeptidase 2; DJ347H13.4; FAM152B; family with sequence similarity 152, member B; Polyubiquitinated substrate transporter; POST; PPPDE peptidase domain containing 2; PPPDE peptidase domain-containing protein 2; PPPDE2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27351 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6ICB0 | |
| DESI1 | |
| synthetic peptide directed towards the C-terminal region of human DESI1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |