Läs mer
Desisobutyl-n-butyl Bortezomib, TRC
Produktkod. 30355375
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30355375 25 mg
1 options
Produktkod. 30355375 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29011525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Desisobutyl-n-butyl Bortezomib
CAS 1104011-35-9
Molekylformel C19H25BN4O4
Kvantitet 25 mg
Synonym B-[(1R)-1-[[(2S)-1-Oxo-3-phenyl-2-[(2-pyrazinylcarbonyl)amino]propyl]amino]pentyl]boronic Acid, Bortezomib N-Pentyl Impurity
InChI formel InChI=1S/C19H25BN4O4/c1-2-3-9-17(20(27)28)24-18(25)15(12-14-7-5-4-6-8-14)23-19(26)16-13-21-10-11-22-16/h4-8,10-11,13,15,17,27-28H,2-3,9,12H2,1H3,(H,23,26)(H,24,25)/t15-,17-/m0/s1
LEDER B([C@H](CCCC)NC(=O)[C@H](CC1=CC=CC=C1)NC(=O)C2=NC=CN=C2)(O)O
Molekylvikt (g/mol) 384.24