Läs mer
Desisobutyryl-ciclesonide, MedChemExpress
Produktkod. 16447197
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16447197 5 mg
16413106 25 mg
16404478 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16447197 Leverantör Medchem Express Leverantörsnummer HY111490
MedChemExpress Desisobutyryl-ciclesonide is the active metabolite of Ciclesonide. Desisobutyryl-ciclesonide has affinity for the glucocorticoid receptor.
Desisobutyryl-ciclesonide is the active metabolite of Ciclesonide. Desisobutyryl-ciclesonide has affinity for the glucocorticoid receptor.
Specifikationer
Kemiskt namn eller material Desisobutyryl-ciclesonide
CAS 161115-59-9
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C28H38O6
Kvantitet 5 mg
Synonym CIC-AP Ciclesonide active principle
Löslighetsinformation DMSO : 100 mg/mL (212.49 mM; Need ultrasonic)
LEDER C[C@@]12[C@@]3(C(CO)=O)[C@@](O[C@H](O3)C4CCCCC4)([H])C[C@@]1([H])[C@]5([H])CCC6=CC(C=C[C@]6(C)[C@@]5([H])[C@@H](O)C2)=O
Molekylvikt (g/mol) 470.6
Formel vikt 470.6
Procent renhet 95.58%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) COVID-19-immunoregulation