Läs mer
Desisobutyryl Ciclesonide, TRC
Produktkod. 30010071
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010071 10 mg
30004976 5 mg
30003305 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010071 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29010010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 161115-59-9
Molekylformel C28 H38 O6
Molekylvikt (g/mol) 470.6
Synonym Pregna-1,4-diene-3,20-dione, 16,17-[[(R)-cyclohexylmethylene]bis(oxy)]-11,21-dihydroxy-, (11β,16α)-, Pregna-1,4-diene-3,20-dione, 16,17-[(cyclohexylmethylene)bis(oxy)]-11,21-dihydroxy-, [11β,16α(R)]-, (11β,16α)-16,17-[[(R)-Cyclohexylmethylene]bis(oxy)]-11,21-dihydroxypregna-1,4-diene-3,20-dione, CIC-AP, Ciclesonide active principle, Desisobutyryl-ciclesonide, (2'R)-2'-Cyclohexyl-11β,21-dihydroxy-16βH-[1,3]dioxolo[4',5':16,17]pregna-1,4-diene-3,20-dione
LEDER C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1C[C@H]5O[C@H](O[C@@]25C(=O)CO)C6CCCCC6
Specifikationer
Kemiskt namn eller material (2'R)-2'-Cyclohexyl-11beta,21-dihydroxy-16betaH-(1,3)dioxolo(4',5':16,17)pregna-1,4-diene-3,20-dione
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C28H38O6/c1-26-11-10-18(30)12-17(26)8-9-19-20-13-23-28(22(32)15-29,27(20,2)14-21(31)24(19)26)34-25(33-23)16-6-4-3-5-7-16/h10-12,16,19-21,23-25,29,31H,3-9,13-15H2,1-2H3/t19-,20-,21-,23+,24+,25+,26-,27-,28+/m0/s1
Formel vikt 470.2668
Procent renhet >95%