Läs mer
Desisopropyl Zilpaterol Hydrochloride, TRC
Produktkod. 30364750
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30364750 10 mg
1 options
Produktkod. 30364750 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29022010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Desisopropyl Zilpaterol Hydrochloride
CAS 92260-83-8
Molekylformel C11 H13 N3 O2 . Cl H
Kvantitet 10 mg
Synonym Imidazo[4,5,1-jk][1]benzazepin-2(1H)-one, 6-amino-4,5,6,7-tetrahydro-7-hydroxy-, hydrochloride (1:1), (6R,7R)-rel-, Imidazo[4,5,1-jk][1]benzazepin-2(1H)-one, 6-amino-4,5,6,7-tetrahydro-7-hydroxy-, monohydrochloride, trans- (9CI), Imidazo[4,5,1-jk][1]benzazepin-2(1H)-one, 6-amino-4,5,6,7-tetrahydro-7-hydroxy-, monohydrochloride, trans-(±)-, Desisopropyl zilpaterol hydrochloride, Desisopropylzilpaterol hydrochloride
InChI formel InChI=1S/C11H13N3O2.ClH/c12-7-4-5-14-9-6(10(7)15)2-1-3-8(9)13-11(14)16;/h1-3,7,10,15H,4-5,12H2,(H,13,16);1H/t7-,10-;/m1./s1
LEDER Cl.N[C@@H]1CCN2C(=O)Nc3cccc([C@H]1O)c23
Molekylvikt (g/mol) 255.7
Formel vikt 255.0775