Läs mer
Desloratadine Epoxide, TRC
Produktkod. 30359986
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30359986 2.5 mg 2.5mg
30353155 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30359986 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2902752.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1346604-23-6
Molekylformel C19H19ClN2O
Molekylvikt (g/mol) 326.82
Synonym Epoxy Desloratadine
LEDER Clc1ccc2c(CCc3cccnc3C24OC45CCNCC5)c1
Specifikationer
Kemiskt namn eller material Desloratadine Epoxide
Kvantitet 2.5 mg
InChI formel InChI=1S/C19H19ClN2O/c20-15-5-6-16-14(12-15)4-3-13-2-1-9-22-17(13)19(16)18(23-19)7-10-21-11-8-18/h1-2,5-6,9,12,21H,3-4,7-8,10-11H2
Formel vikt 326.1186