Läs mer
Desmethoxy Gatifloxacin Trifluoroacetic Acid Salt, TRC
Produktkod. 30354302
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30354302 25 mg
1 options
Produktkod. 30354302 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29045025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Desmethoxy Gatifloxacin Trifluoroacetic Acid Salt
CAS 1215348-50-7
Molekylformel C18H20FN3O3 . C2HF3O2
Kvantitet 25 mg
Synonym 1-Cyclopropyl-6-fluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic Acid Trifluoroacetic Acid Salt, AM 1121Trifluoroacetic Acid Salt, CNV 8912Trifluoroacetic Acid Salt, CNV 97100Trifluoroacetic Acid Salt, ITV 8912Trifluoroacetic Acid Salt
InChI formel InChI=1S/C18H20FN3O3.C2HF3O2/c1-10-8-21(5-4-20-10)16-7-15-12(6-14(16)19)17(23)13(18(24)25)9-22(15)11-2-3-11;3-2(4,5)1(6)7/h6-7,9-11,20H,2-5,8H2,1H3,(H,24,25);(H,6,7)
LEDER CC1CN(CCN1)c2cc3N(C=C(C(=O)O)C(=O)c3cc2F)C4CC4.OC(=O)C(F)(F)F
Molekylvikt (g/mol) 459.4