Läs mer
Desmethyl Cariprazine, TRC
Produktkod. 30009165
Change view
Kvantitet:
1 mg
2.5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30009165 2.5 mg 2.5mg
30005418 1 mg -
30006271 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009165 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2913682.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 839712-15-1
Molekylformel C20H30Cl2N4O
Molekylvikt (g/mol) 413.38
Synonym N-[trans-4-[2-[4-(2,3-Dichlorophenyl)-1-piperazinyl]ethyl]cyclohexyl]-N'-methyl Urea
LEDER ClC1=C(N2CCN(CC[C@@H]3CC[C@@H](NC(NC)=O)CC3)CC2)C=CC=C1Cl
Specifikationer
Kemiskt namn eller material Desmethyl Cariprazine
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C20H30Cl2N4O/c1-23-20(27)24-16-7-5-15(6-8-16)9-10-25-11-13-26(14-12-25)18-4-2-3-17(21)19(18)22/h2-4,15-16H,5-14H2,1H3,(H2,23,24,27)
Procent renhet >95%